Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPR102 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12023R-A488
BOSSBS-12023R-A488EA 893 EUR InStock
BOSSBS-12023R-A488
Anti-GPR102 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Trace amines are endogenous molecules structurally related to classical biogenic amines that are linked to psychiatric conditions. A family of G-protein coupled receptors referred to as trace-amine-associated receptors (TAAR) are activated by trace amines and are present in very low levels in mammalian tissue. TaRs contain several structural features that are similar to the rhodopsin ∫-adrenergic receptor superfamily, including the positions of the seven transmembrane regions that provide common ligand-binding pockets as well as the short N- and C-terminal domains. TAAR proteins are potential targets for drugs of abuse, such as amphetamine and MDMA, as well as neuropsychiatric disorders including schizophrenia, depression, and attention deficit disorder.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR102/TAAR8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge