Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GPCR33 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12030R-A647
BOSSBS-12030R-A647EA 1350 EUR InStock
BOSSBS-12030R-A647
Anti-GPCR33 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
GPRC6A (G protein-coupled receptor family C group 6 member A) is a 928 amino acid protein encoded by the mouse Gprc6a gene. GPRC6A is a member of the G protein-coupled receptor 3 family. This receptor is activated by both amino acids and extracellular concentration of calcium ions. The activity of GPRC6A is mediated by a G protein that activates a phosphatidylinositol-calcium second messenger system. GPRC6A senses changes in the extracellular concentration of calcium ions, suggesting that it may mediate extracellular calcium-sensing responses in osteoblasts. Osteocalin stimulates the activity of GPRC6A in the presence of calcium. GPRC6A also acts as a receptor for amino acids, with a preference for basic amino acids such as L-Lys, L-Arg and L-ornithine. Its affinity for amino acids suggests that it may act as a regulatory component of the urea cycle. GPRC6A is expressed at high levels in liver, lung, spleen and heart, and at lower levels in kidney, skeletal muscle and brain.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPCR6A/GPCR33
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge