Anti-GDE1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-12039R-CY3
BOSSBS-12039R-CY3EA 900 EUR InStock
BOSSBS-12039R-CY3
Anti-GDE1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
GDE1 is a 331 amino acid multi-pass membrane protein that localizes to both the membrane and the cytoplasm and contains one GDPD domain. Expressed in a wide variety of tissues, GDE1 uses magnesium as a cofactor to catalyze the conversion of 1-(sn-glycero-3-phospho)-1D-myo-inositol to myo-inositol and sn-glycerol 3-phosphate, an event that is modulated by G protein signaling pathways and provides a link between phosphoinositide metabolism and G protein signal transduction. The gene encoding GDE1 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MIR16/GDE1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge