Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UNCX Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12043R
BOSSBS-12043REA 725 EUR InStock
BOSSBS-12043R
Anti-UNCX Rabbit Polyclonal Antibody
Antikörper
Members of the paired homeobox family play a role in regulating cell development and pattern formation during embryonic stages. UNCX (UNC homeobox), also known as UNCX4.1, is a 531 amino acid nuclear transcription factor involved in neurogenesis and somitogenesis. Containing one homeobox DNA-binding domain, UNCX belongs to the paired homeobox family and UNC4 subfamily. UNCX assists in the formation of connections between hypothalamic neurons and the pituitary, which is necessary for central neurons to deliver hormones into peripheral blood. UNCX also plays a role in maintaining differentiation of the axial skeleton and acts upstream of Pax-9. The gene encoding UNCX maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UNCX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge