Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Cyclin M3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12045R-A750
BOSSBS-12045R-A750EA 890 EUR InStock
BOSSBS-12045R-A750
Anti-Cyclin M3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Cyclin M3 is a 707 amino acid multi-pass membrane protein that shares weak sequence similarity with cyclin proteins, yet displays no cyclin-like function in vivo. Though ubiquitously expressed, Cyclin M3 is found at highest levels in kidney, brain, spleen and heart. Cyclin M3 is localised to the nucleus where it is likely a metal transporter. Cyclin M3 contains two CBS domains, which appear to bind ligands with an adenosyl group such as AMP, ATP and S-AdoMet and may play a regulatory role in sensitizing proteins to adenosyl-carrying ligands. There are three isoforms of Cyclin M3 that are produced as a result of alternative splicing events.

Type: Primary
Antigen: Cyclin M3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/707
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge