Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-e3K Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12057R-CY7
BOSSBS-12057R-CY7EA 900 EUR InStock
BOSSBS-12057R-CY7
Anti-e3K Rabbit Polyclonal Antibody (Cy7®)
Antikörper
eIF3K (Eukaryotic translation initiation factor 3 subunit K, Muscle-specific gene M9 protein) is a widely expressed translation initiation factor that belongs to the eIF3 subunit K family. Translation initiation factor 3 (eIF3) is a multisubunit complex containing at least 12 subunits. eIF3 binds to the 40S ribosomal subunit, promotes the binding of methionyl-tRNAi and mRNA, and interacts with several other initiation factors to form the 40S initiation complex. eIF3K is the smallest subunit of eIF3 and it interacts with several other subunits of eIF3 and the 40S ribosomal subunit. eIF3K is conserved among high eukaryotes, including mammals, insects, and plants, and it is ubiquitously expressed in human tissues. eIF3K is distributed both in nucleus and cytoplasm and colocalizes with cyclin D3, a regulatory subunit of cyclin-dependent kinase 4 (Cdk4).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: e3K
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge