Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-P2Y10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12070R-A488
BOSSBS-12070R-A488EA 893 EUR InStock
BOSSBS-12070R-A488
Anti-P2Y10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Nucleotides are important extracellular signaling molecules that mediate several events, such as cell proliferation, differentiation, chemotaxis and cytokine release. The P2 receptor family is activated by the binding of nucleotides and is divided into two subfamilies, designated P2X and P2Y. The P2Y receptor family are G protein-coupled receptors that mediate the effects of extracellular nucleotides, primarily through the activation of phospholipase C (PLC). To some extent, the P2Y receptors can also activate potassium channels or, alternatively, inhibit adenylate cyclase and N-type calcium channels in response to extracellular nucleotides. P2Y10 (purinergic receptor P2Y, G-protein coupled, 10), also known as P2RY10, is a 339 amino acid multi-pass membrane protein that is thought to act as a receptor for purines coupled to G-proteins. P2Y10 is found at low levels in blood leukocytes and is upregulated during promyelocytic cell differentiation.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: P2Y10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge