Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-P2Y11 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12071R-FITC
BOSSBS-12071R-FITCEA 900 EUR InStock
BOSSBS-12071R-FITC
Anti-P2Y11 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
P2Y purinoceptor 11 (P2Y11) is a 374 amino acid protein belonging to the G-protein coupled receptor one family. P2Y11 is a multi-pass cell membrane protein that acts as a receptor for both ATP and ATD coupled to G proteins. Due to these interactions, P2Y11 is involved in phosphatidylinositol-calcium and adenylyl cyclase pathways. Induced by DMSO and retinoic acid, P2Y11 is highly expressed in spleen tissue. A putative trans-splicing event involving the gene that encodes P2Y11 and an upstream gene encoding PPAN has been found to result in a fusion protein, designated PPAN-P2RY11.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: P2Y11
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge