Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-P2RY8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12073R-A488
BOSSBS-12073R-A488EA 893 EUR InStock
BOSSBS-12073R-A488
Anti-P2RY8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G-protein coupled receptors translate extracellular signals into intracellular signals (G-protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. P2RY8 (purinergic receptor P2Y, G-protein coupled, 8), also known as P2Y8, is a 359 amino acid multi-pass membrane protein that localizes to the cell membrane and belongs to the G protein-coupled receptor family. Expressed at low levels in lung, heart and kidney, P2RY8 may function as a receptor for purines that are coupled to G proteins and may also play a role in mental retardation.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: P2RY8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge