Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GABRQ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12083R-HRP
BOSSBS-12083R-HRPEA 900 EUR InStock
BOSSBS-12083R-HRP
Anti-GABRQ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Gamma-aminobutyric acid type A (GABAA) receptors mediate inhibitory neurotransmission in the mammalian central nervous system. The receptor exists as a pentameric ion channel composed by heteromeric combinations of alpha, beta, gamma, delta, epsilon, theta, or pi subunits. Only specific subunit combinations produce viable receptors, while others never translocate to the cell surface from the ER where they are synthesized, and subsequently degraded. The theta subunit forms a receptor in combination with alpha3 subunits in monoaminergic cell groups. These receptors, found especially in the septum, preoptic areas, hypothalamic nuclei, amygdala and thalamus, likely have unique pharmacological properties linked to their expression in this particular cell type and not cholinergic cell groups, and may play a role in opiate withdrawl symptoms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GABRQ/GABA A Receptor theta
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge