Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GABRR2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12085R-A350
BOSSBS-12085R-A350EA 893 EUR InStock
BOSSBS-12085R-A350
Anti-GABRR2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Gamma-aminobutyric acid (GABA) receptors are pentameric membrane proteins that operate GABA-gated chloride channels and inhibit neurotransmission in the central nervous system. The rho receptor subunits do not exhibit sensitivity to typical GABA receptor modulators such as bicuculline, hexobarbital, and diazepam. While the rho 1 subunit localizes specifically to the retina, rho 2 expresses in all regions of the brain, though levels were still highest in the retina, implying a role for both subunits in visual pathways.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GABRR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge