Anti-GABRR2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-12085R-A647
BOSSBS-12085R-A647EA 893 EUR InStock
BOSSBS-12085R-A647
Anti-GABRR2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Gamma-aminobutyric acid (GABA) receptors are pentameric membrane proteins that operate GABA-gated chloride channels and inhibit neurotransmission in the central nervous system. The rho receptor subunits do not exhibit sensitivity to typical GABA receptor modulators such as bicuculline, hexobarbital, and diazepam. While the rho 1 subunit localizes specifically to the retina, rho 2 expresses in all regions of the brain, though levels were still highest in the retina, implying a role for both subunits in visual pathways.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: GABRR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge