Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZDHHC7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12088R-A488
BOSSBS-12088R-A488EA 893 EUR InStock
BOSSBS-12088R-A488
Anti-ZDHHC7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZDHHC1 (zinc finger, DHHC-type containing 1), also known as ZNF377 (zinc finger protein 377) or C16orf1, is a 485 amino acid multi-pass membrane protein that contains one DHHC-type zinc finger. Expressed in fetal heart, lung and kidney and also present in adult lung and pancreas, ZDHHC1 functions as a palmitoyltransferase that catalyzes the conversion of palmitoyl-CoA and protein-cysteine to S-palmitoyl protein and CoA. Like ZDHHC1, ZDHHC5, ZDHHC6, ZDHHC7 and ZDHHC18 each contain one DHHC-type zinc finger through which they convey palmitoyltransferase activity against a broad range of substrates, including H-Ras, SNAP 25 and GABAA R proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZDHHC7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge