Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GRID1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12095R-A750
BOSSBS-12095R-A750EA 890 EUR InStock
BOSSBS-12095R-A750
Anti-GRID1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Glutamate receptors mediate most excitatory neurotransmissions in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are divided into two categories, namely NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors consist of seven structurally related subunits, designated GluR-1 to -7, and are primarily responsible for fast excitatory neurotransmissions carried out by glutamate. GluR-delta 1 (Glutamate receptor delta-1 subunit), also known as GRID1, is a multi-pass membrane protein that belongs to the kainate/AMPA receptor family and is expressed primarily in the brain. localised to the cell junction and the postsynaptic cell membrane, GluR-delta 1 functions as a glutamate receptor that regulates synaptic transmissions in the central nervous system (CNS) and is thought to play an important role in synaptic plasticity. Defects in the gene encoding GluR-delta 1 are associated with schizophrenia, a chronic and severe brain disorder.

Type: Primary
Antigen: GRID1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/1009
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge