Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PEX5L Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12096R-A555
BOSSBS-12096R-A555EA 893 EUR InStock
BOSSBS-12096R-A555
Anti-PEX5L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Peroxisomes are single-membrane bound organelles present in virtually all eukaryotic cells. They are involved in numerous catabolic and anabolic pathways, including beta-oxidation of very long chain fatty acids, metabolism of hydrogen peroxide, plasmalogen biosynthesis and bile acid synthesis. The Peroxin gene family, which includes more than 20 members, is required for peroxisome biogenesis. Peroxin 5R, also known as PEX5-related protein or Peroxisome biogenesis factor 5-like, is a 626 amino acid protein that is mainly expressed in brain, with some expression in testis and pancreas. Peroxin 5R contains five TPR repeats, which enable protein-protein interactions and assembly of large multiprotein complexes. There are three isoforms of Peroxin 5R that are produced as a result of alternative splicing events. These isoforms bind C-terminal peroxisome-targeting signals in a similar manner to Peroxin-5. Peroxin 5R interacts with Rab 8b, possibly playing a role in vesicular trafficking and neurotransmitter release.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Peroxin 5R/PEX5-related protein
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge