Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MPDZ Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12099R-A488
BOSSBS-12099R-A488EA 893 EUR InStock
BOSSBS-12099R-A488
Anti-MPDZ Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
MPDZ is a 2,042 amino acid peripheral membrane protein that colocalizes with SR-2C on the apical membrane of epithelial choroid plexus cells. Expressed in heart, brain, placenta, liver, skeletal muscle, kidney and pancreas, MPDZ causes clustering of SR-2C, a serotonin receptor, at the cell surface. MPDZ is member of the NMDAR signaling complex that is involved in regulating AMPAR potentiation and synaptic plasticity in excitatory synapses. As a tight junction protein in epithelial cells, MPDZ interacts with SSTR3, a G-protein-coupled receptor, to regulate transepithelial permeability in a pertussis toxin sensitive manner. MPDZ, along with Kir4.2, may form a complex with other proteins in the nephron and regulate ion transport. MPDZ contains one L27 domain and thirteen PDZ domains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MPDZ
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge