Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NMDAR3A + 3B Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12103R-CY5
BOSSBS-12103R-CY5EA 1350 EUR InStock
BOSSBS-12103R-CY5
Anti-NMDAR3A + 3B Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neuro-degeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors co-localize with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to 7. The kainate/AMPA receptors are primarily responsible for fast excitatory neurotransmission by glutamate, whereas the NMDA receptors exhibit slow kinesis of Ca2+ ions and a high permeability for Ca2+ ions. One such NMDA receptor, NR3B, is expressed in motor neurons and forms cation channels impermeable to calcium, which can resist many open-channel blockers. NR3B functions in the brain as an excitatory glycine receptor, modifying the normal role of glycine as an inhibitory neurotransmitter.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR3A + 3B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge