Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SCN7A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12127R-A555
BOSSBS-12127R-A555EA 893 EUR InStock
BOSSBS-12127R-A555
Anti-SCN7A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Voltage-gated sodium channels are selective ion channels that regulate the permeability of sodium ions in excitable cells. During the propagation of an action potential, sodium channels allow an influx of sodium ions, which rapidly depolarize the cell. The three glycoproteins that comprise the voltage-gated sodium channel proteins include a pore-forming Beta subunit, a noncovalently associated Beta 1 subunit and a disulfide-linked Beta 2 subunit. Na+ CP type VII Alpha (Sodium channel protein type 7 subunit alpha), also known as SCN6A, Sodium channel protein cardiac and skeletal muscle subunit alpha and putative voltage-gated sodium channel subunit alpha Nax, is a 1682 amino acid multi-pass membrane protein that belongs to the sodium channel family. Primarily expressed in uterus and heart, Na+ CP type VII Alpha may function in the regulation of salt intake behavior and central sensing of body-fluid sodium levels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCN7A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge