Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ACCN2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12128R-CY7
BOSSBS-12128R-CY7EA 900 EUR InStock
BOSSBS-12128R-CY7
Anti-ACCN2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Acid sensing ion channel ASIC1 is present in brain as a 4.3-kb transcript with localization to rat dorsal root ganglia. In situ hybridization of rat brain suggests that ASIC1 is most abundant in the main olfactory bulb, cerebral cortex, hippocampal formation, habenula, basolateral amygdaloid nuclei and cerebellum. ASIC1 and H+-gated currents may contribute to the development of fear and anxiety. ASIC2, also designated amiloride-sensitive cation channel 1, neuronal (ACCN1), mammalian degenerin, BNAC1 (MDEG) and brain Na+ channel 1, mediates the normal detection of light touch. ASIC2 mRNA is abundant in brain, specifically in neurons. ASIC2 is expressed as 2.7- and 3.7-kb transcripts in brain and spinal cord tissues. ASIC3, also designated SLNAC1 and TNaC1, mediates detection of lasting pH changes and is involved in modulating moderate- to high-intensity pain sensation. ASIC4, also designated ACCN4 and BNAC4, is abundant in pituitary gland and is also present in the inner ear.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACCN2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge