Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PNCK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12134R-HRP
BOSSBS-12134R-HRPEA 900 EUR InStock
BOSSBS-12134R-HRP
Anti-PNCK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The Ca2+/calmodulin-dependent protein kinases (CaMKs) comprise a structurally related subfamily of serine/threonine kinases. CaMKI Beta (Ca2+/calmodulin-dependent protein kinase type 1B), also known as PNCK (pregnancy up-regulated non-ubiquitously expressed CaM kinase) or BSTK3, is a 343 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one protein kinase domain. Existing as multiple alternatively spliced isoforms, CaMKI Beta functions to catalyze the ATP-dependent phosphorylation of CaMKI, an event that activates CaMKI activity and may be important for Ca2+-triggered signaling cascades within the cell. The gene encoding CaMKI Beta maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PNCK
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge