Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-InaD-Like Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12142R-A647
BOSSBS-12142R-A647EA 893 EUR InStock
BOSSBS-12142R-A647
Anti-InaD-Like Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The membranes of myelinating Schwann cells are joined by tight, gap and adherens junctions, all of which are found in regions of noncompact myelin: the paranodal loops, incisures of Schmidt-Lanterman and mesaxons. Tight junctions help establish polarity in mammalian epithelia by forming a physical barrier that separates apical and basolateral membranes. Pals-associated tight junction protein (PATJ), the human homolog of Drosophila Discs Lost, is differentially localized in myelinating Schwann cells. PATJ associates with Claudin-1, CRB1 (a transmembrane protein that plays a role in epithelial cell polarity and photoreceptor development), and Pals1 (a Lin-7 associated protein). The PATJ/Pals1/CRB1 complex can form a tripartite tight junction in epithelial cells crucial to their integrity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: InaD-Like
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge