Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DBX2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12149R-CY5
BOSSBS-12149R-CY5EA 900 EUR InStock
BOSSBS-12149R-CY5
Anti-DBX2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
DBX2 is a 339 amino acid member of the H2.0 homeobox family. DBX2, which is localized to the nucleus, contains one homeobox DNA-binding domain, a region of 60 amino acids that binds DNA through a helix-turn-helix type of structure. DBX2, which is expressed in the forebrain, midbrain, hindbrain and spinal cord, has been implicated in CNS development.Specifically, DBX2 has been shown to play a role in spinal cord dorsal/ventral patterning, as well as the regionalization of the CNS. DBX2 is also thought to play a role in the production of multiple spinal cord cell types.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DBX2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge