Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TXLNB Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12153R-CY7
BOSSBS-12153R-CY7EA 900 EUR InStock
BOSSBS-12153R-CY7
Anti-TXLNB Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Beta-taxilin promotes nerve regeneration and may be involved in intracellular vesicle transport. Expressed predominantly in cardiac and skeletal muscle, beta-taxilin binds to the coiled coil region of the syntaxin family members STX1A, STX3A, and STX4A. Beta- and gamma-taxilins, bind to the alpha subunit of the nascent polypeptide-associated complex (NAC) and affect its nuclear distribution, suggesting that the taxilin family is involved not only in the translational process through its interaction with NAC but also in the transcriptional process through its interaction with alphaNAC alone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TXLNB
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge