Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAPS1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12161R-CY3
BOSSBS-12161R-CY3EA 900 EUR InStock
BOSSBS-12161R-CY3
Anti-CAPS1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CAPS1 is a 189 amino acid cytoplasmic protein that contains four EF-hand domains, which serve as calcium-binding sites, and was first identified in canine thyroid. Interestingly, CAPS1 is much less abundant in humans than in canines. Synthesis and phosphorylation of CAPS1 is upregulated by cAMP-agonists in thyrocytes. CAPS1 likely functions in the regulation of ionic transport and may be involved in cross-signaling between cAMP and Ca(+2)-phophatidylinositol cascades. In addition to thyriod, CAPS1 is expressed in brain, salivary glands and lung. Expression of CAPS1 is increased in endometrial cancer and prognosis seems to be dependent on the level of CAPS1 expression, indicating that CAPS1 may be an appropriate prognostic marker for patient survival.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAPS1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge