Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Calcyphosine 2/CAPS2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12163R-A680
BOSSBS-12163R-A680EA 890 EUR InStock
BOSSBS-12163R-A680
Anti-Calcyphosine 2/CAPS2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
CAPS2 is a 557 amino acid calcium-binding protein that is abundantly expressed, with highest expression found in placenta, testis, colon, lung and brain. CAPS2 contains three EF-hand domains and exists as three alternatively spliced isoforms. Suggested to play a role in large dense-core vesicle (LDCV) exocytosis, CAPS2 is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.

Type: Primary
Antigen: Calcyphosine 2/CAPS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 221-320/557
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge