Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FBN2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12166R-CY5
BOSSBS-12166R-CY5EA 900 EUR InStock
BOSSBS-12166R-CY5
Anti-FBN2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Extracellular glycoproteins fibrillin-1 and -2 are major components of connective tissue microfibrils. Fibrillin-2 containing microfibrils regulate the early process of elastic fiber assembly in tissue. Mutations in the fibrillin-2 gene resulting in impaired assembly of fibrillin-2 may lead to molecular congenital contractural arachnodactyly. Fibrillin-2 constitutes the backbone of microfibrils which insert directly into the lamina densa of basement membranes. Epithelial cells primarily deposit fibrillin into the extracellular matrix in a nonfibrillar form. Mutations in the 8-cysteine motif of Fibrillin-2 alters its binding to microfibril-associated glycoprotein-1 (MAGP-1), which may increase the severity of congenital contractural arachnodactyly.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fibrillin 2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge