Anti-KCNF1 Rabbit Polyclonal Antibody
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-12173R
BOSSBS-12173REA 704 EUR InStock
BOSSBS-12173R
Anti-KCNF1 Rabbit Polyclonal Antibody
Antikörper
KCNF1 is a multi-pass membrane-bound protein that acts as an ion channel and is generally expressed as a heterotetramer of potassium channeling proteins. Formerly known as kH1, KCNF1 is usually found as a heteromer with three other potassium channel proteins, KCNG3, KV6.3 and KCNV2. As a potassium channel protein, KCNF1 plays a role in regulating apoptosis and proliferation of pulmonary artery smooth muscle (PASM) cells. Bone morphogenetic proteins (BMPs) restrict proliferation and can induce apoptosis in normal human PASM cells and will upregulate expression of KCNF1 in PASM cells in vitro. KCNF1 is expressed in heart, brain, liver, skeletal muscle, kidney and pancreas.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: KCNF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge