Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KCNA2B Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12182R-CY5
BOSSBS-12182R-CY5EA 900 EUR InStock
BOSSBS-12182R-CY5
Anti-KCNA2B Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles, and other excitable cells. The KV gene family encodes more than 30 genes that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution, and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming a-subunits (KV), which include the KV1, KV2, KV3, and KV4 proteins, and accessory or KV-subunits that modify the gating properties of the coexpressed KV subunits. Differences exist in the patterns of trafficking, biosynthetic processing, and surface expression of the major KV1 subunits (KV1.1, KV1.2, and KV1.4) expressed in rat and human brain, suggesting that the individual protein subunits are highly regulated to control for the assembly and formation of functional neuronal channels. KV beta.2 can also be designated KCNAB2, KKv beta2.1 or AKR6A5.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KCNA2B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge