Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DZIP3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12191R-HRP
BOSSBS-12191R-HRPEA 900 EUR InStock
BOSSBS-12191R-HRP
Anti-DZIP3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
DZIP3 is a 1,208 amino acid protein that localizes to the cytoplasm and contains one RING-type zinc finger. Expressed in a variety of tissues with highest expression in heart, skeletal muscle and kidney, DZIP3 functions as an E3 ubiquitin-protein ligase that accepts ubiquitin from an E2 ubiquitin-conjugating enzyme, thereby playing a role in signaling events throughout the cell. Multiple isoforms of DZIP3 exist due to alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DZIP3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge