Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FOXD1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12193R-CY7
BOSSBS-12193R-CY7EA 900 EUR InStock
BOSSBS-12193R-CY7
Anti-FOXD1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Transcription factor required for formation of positional identity in the developing retina, regionalization of the optic chiasm and morphogenesis of the kidney. Can neuralize ectodermal cells directly By similarity. Binds to the consensus sequence 5'-A[AT]T[AG]TTTGTTT-3' and acts as a transcriptional repressor. Also acts as a transcriptional activator. Promotes development of neural crest cells from neural tube progenitors. Restricts neural progenitor cells to the neural crest lineage while suppressing interneuron differentiation. Required for maintenance of pluripotent cells in the pre-implantation and peri-implantation stages of embryogenesis. Probable transcription factor involved in embryogenesis and somatogenesis. FOXD1 is involved in regulating inflammation as well as kidney and retinal development. FOXD1 regulates the activity of NFAT and NFkB. Deficiency of FOXD1 results in multiorgan systemic inflammation, exaggerated Th cell-derived cytokine production, and T cell proliferation in autogolgous MLRs. In kidneys, FOXD1 controls the production of signals required for the normal transition of induced mesenchyme into tubular epithelium and full growth and branching of the collecting system. Deletion of FOXD1 results in renal abnormalities. FOXD2 acts as a modulator of T cell activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXD
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge