Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF350 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12213R-CY5
BOSSBS-12213R-CY5EA 900 EUR InStock
BOSSBS-12213R-CY5
Anti-ZNF350 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The transcpritional repressor ZBRK1, also designated zinc finger and BRCA1-interacting protein with a KRAB domain 1 or zinc finger protein 350, belongs to the Krüppel C2H2-type zinc-finger protein family of proteins. ZBRK1 localizes mainly to the nucleus and may be associated with the nuclear matrix. It is a widely expressed protein that binds to BRCA1. ZBRK1 plays an important role in transcriptional regulation. Likely targets of gene regulation are DNA damage response genes, which effect the survival and growth control of cells. ZBRK1 contains an N-terminal KRAB domain, a C-terminal BRCA1-binding region and eight central zinc-fingers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF350
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge