Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF426 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12219R-CY7
BOSSBS-12219R-CY7EA 900 EUR InStock
BOSSBS-12219R-CY7
Anti-ZNF426 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF426 (Zinc finger protein 426), also known as MGC2663, is a 554 amino acid protein that is thought to be involved in transcriptional regulation. Localized to the nucleus, ZNF426 contains one KRAB domain and 12 C2H2-type zinc fingers through which it may convey DNA, RNA and protein binding capabilities. Specifically, ZNF426 may interact with the viral protein KSHV ORF 50 and, through this interaction, may activate viral gene transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF426
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge