Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZNF750 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12239R-A555
BOSSBS-12239R-A555EA 893 EUR InStock
BOSSBS-12239R-A555
Anti-ZNF750 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 750 is a 723 amino acid member of the Krüppel C2H2-type zinc finger protein family. Localized to the nucleus, ZNF750 contains one conserved C2H2 zinc finger domain and is expressed in the skin, lungs, prostate, placenta and thymus. ZNF750 is also expressed in primary human keratinocytes but not in fibroblasts. Mutations in the gene encoding ZNF750 cause Seborrhea-like dermatitis with psoriasiform, a condition characterized by a chronic and diffuse rash on the face and hyperkeratosis of skin over the elbows, soles, knees and palms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF750
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge