Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PUM2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12246R-FITC
BOSSBS-12246R-FITCEA 900 EUR InStock
BOSSBS-12246R-FITC
Anti-PUM2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Pumilio 2 is a sequence-specific RNA-binding protein that regulates translation and mRNA stability by binding mRNA targets. It supports proliferation and self-renewal of stem cells by regulating the translation of key transcripts. The Pumilio gene encodes proteins that are required for development of germ stem cells in one or both sexes. The Pumilio protein interacts with the human Nanos1 protein and this interaction may play a conserved role in germ cell development. Pumilio 2 is highly expressed in testis and ovary and at lower levels in brain, heart, kidney, liver, muscle, placenta, intestine and stomach. It is also expressed in stem cells, germ cells and in most fetal tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PUM2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge