Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ZFP36L1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12254R-A350
BOSSBS-12254R-A350EA 893 EUR InStock
BOSSBS-12254R-A350
Anti-ZFP36L1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
TIS11B is a member of the tristetraprolin family. Tristetraprolin (TTP), or TIS11, is a zinc-binding protein encoded by the immediate-early response gene, Zfp-36. TIS11B, a relative of TTP, localizes to the nucleus and may function as a transcription factor involved in regulating the growth factor response. It is an evolutionarily conserved protein containing two C3H1-type zinc fingers and a repeating cys-his motif. TIS11B is an mRNA binding protein and is known to interact with the 3’-untranslated region of VEGF mRNA, thereby decreasing its stability. This suggests that TIS11B is a potential target in antiangiogenic therapy. In addition, TIS11B may also be an important regulator of myogenesis, as its expression is upregulated during murine myoblast differentiation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZFP36L1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge