Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FUT11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12259R-A647
BOSSBS-12259R-A647EA 893 EUR InStock
BOSSBS-12259R-A647
Anti-FUT11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which are essential interfaces for biological recognition processes. Fucosyltransferases (FucTs) catalyze the covalent association of fucose to different positional linkages on sugar acceptor molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. FucT-XI is a 492 amino acid single-pass type II membrane protein that belongs to the glycosyltransferase 10 family. Localizing to Golgi apparatus, FucT-XI may act as a fucosyltransferase and exists as two alternatively spliced isoforms. The gene encoding FucT-XI maps to mouse chromosome 14 A3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FUT11/FucT-XI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge