Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RTF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12261R-A680
BOSSBS-12261R-A680EA 890 EUR InStock
BOSSBS-12261R-A680
Anti-RTF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
RTF1 is the the major histocompatibility complex (MHC) in the rat which contains genes that code for two class II histocompatibility antigens. RT1-B is an antigen of the RT1 complex. It is a protein dimer consisting of an alpha and beta glycoprotein chain and is homologous to I-A and I-E genes, respectively, in the H-2 complex of the mouse. MHC Class II antigens are useful in studying T helper cell interaction with class II positive antigen presenting cells (dendritic cells, B cells, macrophages) and offer new possibilities for studying the development of T helper cells since these also stain stromal cells in the thymus.

Type: Primary
Antigen: RTF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 231-350/710
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge