Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GGNBP2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12267R
BOSSBS-12267REA 1090 EUR InStock
BOSSBS-12267R
Anti-GGNBP2 Rabbit Polyclonal Antibody
Antikörper
LCRG1 is a 697 amino acid protein that may be involved in spermatogenesis. LCRG1 interacts with gametogenetin, a protein primarily expressed in testis and ovary. Localized in the cytoplasm, LCRG associates with vesicular structures. LCRG1 is expressed in brain, placenta, lung, liver and kidney, with highest levels found in heart, pancreas and skeletal muscle. LCRG1 is strongly down-regulated in 40% of primary laryngeal carcinoma and in 6 of 10 various cancer cell lines. Phosphorylated on Serine 360, LCRG1 exists as three isoforms produced by alternative splicing events.

Recommended Dilutions: IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: GGNBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge