Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HOOK1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12287R-A350
BOSSBS-12287R-A350EA 876 EUR InStock
BOSSBS-12287R-A350
Anti-HOOK1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Microtubules mediate the spatial organization of diverse membrane-trafficking systems. The HOOK proteins, HOOK1, HOOK2 and HOOK3, comprise a family of cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules; and more divergent C-terminal domains, which mediate binding to organelles. HOOK1, a cytoskeletal linker protein, may play a role in endocytic membrane trafficking. It exists as a homodimer, most likely mediated through its central coiled-coil domain. HOOK1 interacts with VPS18 and is required for spermatid differentiation, in which it is most likely involved in the positioning of the manchette microtubules and the flagellum. HOOK1 localizes primarily to the cytoplasm and does not associate with the Golgi complex, unlike HOOK3, which participates in the organization of the cis-Golgi compartment.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HOOK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge