Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TESK2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12293R-A488
BOSSBS-12293R-A488EA 893 EUR InStock
BOSSBS-12293R-A488
Anti-TESK2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
TESK2 is a nuclear protein that belongs to the protein kinase superfamily and is expressed in testis and prostate. Functioning as a dual-specificity protein kinase, TESK2 catalyzes the ATP-dependent phosphorylation of substrates and autophosphorylation on tyrosine and serine/threonine residues, thereby mediating intracellular signal transduction pathways. TESK2 requires magnesium as a cofactor and its catalytic activity is thought to play an important role in meiotic events such as spermatogenesis. TESK2 contains one protein kinase domain that is 65% identical to the kinase domain found in TESK1 (testicular protein kinase 1), suggesting a similar role for these proteins in phosphorylation events. Three isoforms of TESK2 are expressed due to alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TESK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge