Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HEMGN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12303R-HRP
BOSSBS-12303R-HRPEA 900 EUR InStock
BOSSBS-12303R-HRP
Anti-HEMGN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Hemogen is a 484 amino acid protein encoded by the human gene HEMGN. Hemogen is a nuclear protein that is expressed in hematopoietic precursor cells and can be detected in CD34+ and K-562 leukemia cell line. It is also expressed in bone marrow, testis, thymus and thyroid tumors, non-Hodgkin lymphoma, various leukemia cell lines, peripheral blood mononuclear cells (PBMCs) and bone marrow mononuclear cells (BMMCs) of patients with leukemia. Hemogen is down-regulated during megakaryocytic differentiation of K-562 cells by 12-O-tetradecanoylphorbol-13-acetate (TPA) (at protein level). It can be up-regulated in normal PBMCs by mitogens.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: HEMGN
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge