Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-USE1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12305R-CY3
BOSSBS-12305R-CY3EA 900 EUR InStock
BOSSBS-12305R-CY3
Anti-USE1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
In eukaryotic cells, the Golgi apparatus receives newly synthesized proteins from the endoplasmic reticulum (ER) and, after covalent modification, delivers them to their destination in the cell. For membrane-directed proteins this process is believed to be carried out via vesicular transport. Correct vesicular transport is determined by specific pairing of vesicle-associated SNAREs (v-SNAREs) with those on the target membrane (t-SNAREs). Unconventional SNARE in the ER 1, also known as USE1 or protein p31, is a 259 amino acid t-SNARE that forms a larger complex with ZW10, RINT-1 and Syntaxin 18. Upon Mg2+-AP treatment in the presence of NSF and ?SNAP, ZW10, RINT-1 and USE1 dissociate from Syntaxin 18. USE1 is a single-pass type IV membrane protein that is localized to the endoplasmic reticulum membrane. Three named isoforms exist for USE1 as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: USE1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge