Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PHEX Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12313R-HRP
BOSSBS-12313R-HRPEA 900 EUR InStock
BOSSBS-12313R-HRP
Anti-PHEX Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The PHEX a 749 amino acid protein that putatively consists of an intracellular, transmembrane and extracellular domain. PHEX mutations have been observed in 60-80% of hypophosphatemic rickets patients. The PHEX protein, which is a single-pass membrane protein, is also designated HYP, X-linked hypophosphatemia protein or metalloendopeptidase homolog PEX. PHEX plays an active role in bone and dentin mineralization and renal phosphate re-absorption. X-linked hypophosphatemic rickets, also designated HYP, is an X-linked dominant disorder characterized by impaired phosphate uptake in the kidney, which is likely to be caused by abnormal regulation of sodium phosphate cotransport in the proximal tubules. Clinical manifestations include skeletal deformities, growth failure, craniosynostosis, paravertebral calcifications, pseudofractures in lower extremities, and muscular hypotonia with onset in early childhood.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PHEX
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge