Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HHATL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12315R-FITC
BOSSBS-12315R-FITCEA 900 EUR InStock
BOSSBS-12315R-FITC
Anti-HHATL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
GUP1 is a 504 amino acid multipass membrane protein of the endoplasmic reticulum that functions as a membrane bound O-acyltransferase. With specific expression in heart, GUP1 negatively regulates amino-terminal palmitoylation of Shh by HHAT, a protein that is required for Shh signaling. Deletion of the gene encoding GUP1 results in higher sensibility to specific sphinogolipid biosynthesis inhibitors and resistance to ergosterol biosynthesis inhibitors, indicating that GUP1 is an essential component in lipid metabolism. Also, GUP1 also seems to be important for cell wall assembly and stability due to evidence in Saccharomyces cerevisiae GUP1 mutants, which exhibit altered plasma membrane lipid composition and membrane potential.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HHATL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge