Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SCUBE3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12317R-CY5
BOSSBS-12317R-CY5EA 900 EUR InStock
BOSSBS-12317R-CY5
Anti-SCUBE3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
SCUBE3 is a novel secreted 993 amino acid cell-surface osteoblast protein that plays an important role in bone cell biology. While highly expressed in osteoblasts, SCUBE3 expression is unobservable or very low in non-bone tissues. SCUBE3 forms homo-oligomers and hetero-oligomers with SCUBE1, and may undergo C-terminal proteolytic cleavage or become N-glycosylated following translation. Two SCUBE3 isoforms exist as a result of alternative splicing events. SCUBE3 contains one CUB domain and nine EGF-like domains, and is encoded by a gene which maps to human chromosome 6p21.31, a region associated with a rare form of metabolic bone disease known as Paget's disease. Chromosome 6 contains 170 million base pairs and comprises nearly 6% of the human genome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCUBE3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge