Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LY6C Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12327R-A488
BOSSBS-12327R-A488EA 876 EUR InStock
BOSSBS-12327R-A488
Anti-LY6C Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The gene encoding the mouse alloantigen, Ly-6C, maps to chromosome 15 and encodes a 131 amino acid protein that belongs to the Ly-6 family of glycosyl-phosphatidylinositol (GPI)-linked proteins. Ly-6 family members share amino acid homology throughout a distinctive cysteine rich protein domain that incorporates O-linked carbohydrates. Murine Ly-6 molecules have unique patterns of tissue expression during hematopoiesis from multipotential stem cells to lineage committed precursor cells, and on specific leukocyte subpopulations in the peripheral lymphoid tissues. Ly-6C is predominantly expressed on murine peripheral CD8 T cells. Ly-6C is involved in endothelial adhesion, the killing of target cells by CTLs, inducing TCR-mediated activation of IL-2 and IFN-?production in CD8 T cells and the homing of CD8 T cells. In addition, Ly-6C may act as a signaling molecule of LFA-1 activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LY6C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge