Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Trim22/Staf-50 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12330R-A680
BOSSBS-12330R-A680EA 890 EUR InStock
BOSSBS-12330R-A680
Anti-Trim22/Staf-50 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The tripartite motif (TRIM) family of proteins are characterised by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. Staf-50 (50 kDa-stimulated trans-acting factor), also known as TRIM22 (tripartite motif-containing 22), RNF94 or GPSTAF50, is a 498 amino acid cytoplasmic protein that belongs to the TRIM family and, characteristic of TRIM family members, contains one RING-type zinc finger, one B box-type zinc finger and one SPRY domain. Induced by IFN-å and IFN-, Staf-50 is strongly expressed in ovary, spleen, thymus and peripheral blood leukocytes where it is thought to mediate the antiviral effects of IFN proteins. Additionally, Staf-50 is present in leukemic cells, suggesting a role in cancer formation and metastasis. Staf-50 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 11.

Type: Primary
Antigen: Trim22/Staf-50
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 231-330/498
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge