Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ABCB7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12331R-A647
BOSSBS-12331R-A647EA 893 EUR InStock
BOSSBS-12331R-A647
Anti-ABCB7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The peroxisomal membrane contains several ATP-binding cassette (ABC) transporters, ABCD1–4 that are known to be present in the human peroxisome membrane (1). All four proteins are ABC half-transporters, which dimerize to form an active transporter (1). A mutation in the ABCD1 causes X-linked adrenoleukodystrophy (X-ALD), a peroxisomal disorder which affects lipid storage (2,3). ABCD2 in mouse, is expressed at high levels in the brain and adrenal organs, which are adversely affected in X-ALD (4). The peroxisomal membrane comprises 2 quantitatively major proteins, PMP22 and ABCD3 (5). ABCD3 is associated with irregularly shaped vesicles which may be defective peroxisomes or peroxisome precursors (5). ABCD4 localizes to peroxisomes (1). The genes which encode ABCD1–4 map to human chromosome Xq28, 12q11-q12, 1p22-p21 and 14q24.3, respectively (3,6–8). ABCB7 is a half-transporter involved in the transport of heme from the mitochondria to the cytosol and maps to human chromosome Xq13.1-q13.3 (9).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCB7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge