Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DYRK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12332R-FITC
BOSSBS-12332R-FITCEA 900 EUR InStock
BOSSBS-12332R-FITC
Anti-DYRK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Dyrk is the homolog of the Drosophila mnb (minibrain) gene, which is required for neurogenesis (1–3). Dyrk is a dual-specificity tyrosine kinase and serine/threonine kinase, which is itself regulated by tyrosine phosphorylation (1). Several mammalian Dyrk related proteins have been identified and are thought to compose a family of dual specificity protein kinases (4). Dyrk family members, including Dyrk1A (originally Dyrk), Dyrk1B, Dryk1C, Dyrk2, Dyrk3, Dyrk4A and Dyrk4B, are thought to be involved in diverse cellular functions (4). Dyrk1A is a candidate gene that may be involved in Downs syndrome, and it has been found to be somewhat overexpressed in Downs syndrome (1,5). Two isoforms of human fetal brain Dyrk2 exist: a deduced 528-amino acid protein and a protein containing 73 additional amino acids at the amino terminus (4). Dyrk3 is strongly expressed in testis, only after the onset of spermatogenesis, and very weakly expressed in spleen and adrenal gland (1). The genes which encode Dyrk2 and Dyrk3 map to human chromosomes 12 and 1q32, respectively (4).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DYRK2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge