Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ERMAP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-12333R-A555
BOSSBS-12333R-A555EA 893 EUR InStock
BOSSBS-12333R-A555
Anti-ERMAP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ERMAP is a single-pass type one membrane protein that belongs to the immunoglobulin superfamily. Expressed in cord blood, fetal liver and adult bone marrow, ERMAP is thought to function as a cell adhesion molecule in erythroid cells and is responsible for expression of the Scianna/Radin (Sc/Rd) blood group antigen system. The Sc/Rd system is comprised of seven antigens that are present on the surface of red blood cells and have a variety of functions ranging from protein transport to cell adhesion. These seven blood antigens can differ in their expression within a population and may sometimes differ between mother and child. A fetus expressing different blood antigens than its mother may cause the mother to produce against the fetal blood. This condition is known as hemolytic disease of the newborn (HDN) and is characterized by jaundice, anemia and in some cases, infant death.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ERMAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge